3-Phenyl-2-propyn-1-ol ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Phenylprop-2-yn-1-ol

136975-25G 136975-5G
BT136975-25GEA 209 USD
BT136975-25G BT136975-5G
3-Phenyl-2-propyn-1-ol ≥98%
3-Phenyl-2-propyn-1-ol

CAS: 1504-58-1; MDL No: MFCD00040914 Liquid; Linear Formula: C6H5C≡CCH2OH; Molecular Formula: C9H8O; MW: 132.16 Boiling Point: 129-130°/10 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.06; Refractive Index: 1.585


  • Refractive index: 1.585
  • Liquid

Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 129…130 °C/10 mmHg
Density: 1.06 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00040914
CAS Number: 1504-58-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR