2,3-Dichloroisonicotinic acid ≥98%
Supplier: BeanTown Chemical
136965-1G
136965-5G
BT136965-1GEA
94
USD
BT136965-1G
BT136965-5G
2,3-Dichloroisonicotinic acid ≥98%
2,3-Dichloroisonicotinic acid
CAS: 184416-84-0
Solid; Molecular Formula: C6H3Cl2NO2; MW: 192.00
Melting Point: 203-207°
- Solid
Formula:
C₆H₃Cl₂NO₂ Melting Pt: 203…207 °C |
MDL Number:
MFCD01861977 CAS Number: 184416-84-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...