(-)-Dibenzoyl-L-tartaric acid, anhydrous ≥99%

Supplier: BeanTown Chemical

Synonyms: (2R,3R)-2,3-Bis(benzoyloxy)succinic acid

136920-100G 136920-25G 136920-500G
BT136920-100GEA 74 USD
BT136920-100G BT136920-25G BT136920-500G
(-)-Dibenzoyl-L-tartaric acid, anhydrous ≥99%
(-)-Dibenzoyl-L-tartaric acid

CAS: 2743-38-6; EC No: 220-374-0; MDL No: MFCD00003074; RTECS: WW8070000 Powder; Molecular Formula: C18H14O8 ; MW: 358.30 Melting Point: 154-156° Optical Rotation: [α]20/D −116°, c = 9 in ethanol


  • RTECS: WW8070000
  • Optical rotation: [α]20/D −116°, c = 9 in ethanol
  • Powder

Formula: C₁₈H₁₄O₈
MW: 358.31 g/mol
Melting Pt: 154…156 °C
MDL Number: MFCD00003074
CAS Number: 2743-38-6
EINECS: 220-374-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR