Tetrahydro-4H-pyran-4-one ≥98%

Supplier: BeanTown Chemical

Synonyms: Dihydro-2H-pyran-4(3H)-one, Tetrahydro-α-pyran-4-one, Tetrahydro-2H-pyran-4-one, Tetrahydro-γ-pyran-4-one

136890-1G 136890-25G 136890-5G
BT136890-1GEA 46 USD
BT136890-1G BT136890-25G BT136890-5G
Tetrahydro-4H-pyran-4-one ≥98%
Tetrahydro-4H-pyran-4-one

CAS: 29943-42-8; EC No: 249-967-2; MDL No: MFCD00006581 Liquid; Molecular Formula: C5H8O2; MW: 100.12 Boiling Point: 166-167°; Flash point: 55°C (131°F) Density (g/mL): 1.084; Refractive Index: 1.452


  • Refractive index: 1.452
  • Liquid

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 166…167 °C
Density: 1.084 g/mL
Flash Pt: 55 °C (131 °F)
MDL Number: MFCD00006581
CAS Number: 29943-42-8
EINECS: 249-967-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR