5-Fluorocytosine ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one

136875-1G 136875-250MG
BT136875-1GEA 93.39 USD
BT136875-1G BT136875-250MG
5-Fluorocytosine ≥98%
5-Fluorocytosine

CAS: 2022-85-7; EC No: 217-968-7; MDL No: MFCD00006035; RTECS: HA6040000 Powder; Molecular Formula: C4H4FN3O; MW: 129.09 Melting Point: 298-300° (decomposes) Light Sensitive


  • RTECS: HA6040000
  • Light Sensitive
  • Powder

Formula: C₄H₄FN₃O
MW: 129.09 g/mol
Boiling Pt: 298
Melting Pt: 165 (dec.)
MDL Number: MFCD00006035
CAS Number: 2022-85-7
EINECS: 217-968-7
Merck Index: 13,04151

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR