Coomassie brilliant blue G-250
Supplier: BeanTown Chemical
Synonyms:
C.I. 42655
136865-25G
136865-5G
BT136865-25GEA
123
USD
BT136865-25G
BT136865-5G
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250
CAS: 6104-58-1; EC No: 228-058-4; MDL No: MFCD00078482
Powder; Molecular Formula: C47H48N3NaO7S2; MW: 854.02
- Powder
Formula:
C₄₇H₄₈N₃NaO₇S₂ MW: 854.04 g/mol |
MDL Number:
MFCD00078482 CAS Number: 6104-58-1 EINECS: 228-058-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...