3-Chlorophenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Chlorobenzeneboronic acid, (3-Chlorophenyl)metaboric acid, 3-Chlorophenyl boronic acid, Dihydroxy(3-chlorophenyl)borane

136855-1G 136855-25G 136855-5G
BT136855-1GEA 22.97 USD
BT136855-1G BT136855-25G BT136855-5G
3-Chlorophenylboronic acid ≥97%
3-Chlorophenylboronic acid

CAS: 63503-60-6; MDL No: MFCD00161354 Crystalline/Powder; Molecular Formula: C6H6BClO2; MW: 156.37 Melting Point: 185-189°


  • Crystalline/Powder

Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 185…189 °C
MDL Number: MFCD00161354
CAS Number: 63503-60-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR