Sodium tetrachloropalladate(II) hydrate ≥99.95% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Palladium sodium chloride, Sodium palladium (II) chloride, Disodium tetrachloropalladate

136835-1G 136835-5G
BT136835-1GEA 75.47 USD
BT136835-1G BT136835-5G
Sodium tetrachloropalladate(II) hydrate ≥99.95% (trace metals basis)
Sodium tetrachloropalladate(II) hydrate

CAS: 13820-53-6; EC No: 237-502-6; MDL No: MFCD00003487; RTECS: RT3480000 Crystalline/Powder; Linear Formula: Na2PdCl4Na2·xH2O; MW: 294.20 (anhydrous) Hygroscopic

Formula: Cl₄Na₂Pd·xH₂O
MW: 294.21 g/mol (anhy)
MDL Number: MFCD00003487
CAS Number: 13820-53-6

Order Now

Specification Test Results

Assay 99.95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR