Trifluoroacetaldehyde hydrate ≥75%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Trifluoroacetaldehyde hydrate, 2,2,2-Trifluoro-1,1-ethanediol
136825-10G
136825-50G
BT136825-10GEA
90
USD
BT136825-10G
BT136825-50G
Trifluoroacetaldehyde hydrate ≥75%, Technical Grade
Trifluoroacetaldehyde hydrate
CAS: 421-53-4; EC No: 207-006-4; MDL No: MFCD00167359; RTECS: KI2850000
Liquid; Molecular Formula: C2H3F3O2; MW: 116.04 (anhydrous)
Boiling Point: 104-106°
Density (g/mL): 1.44
- RTECS: KI2850000
- Liquid
Formula:
C₂H₃F₃O₂ MW: 116.039 g/mol Boiling Pt: 104…106 °C Density: 1.44 g/mL |
MDL Number:
MFCD00167359 CAS Number: 421-53-4 EINECS: 207-006-4 |
Specification Test Results
Assay | 75% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...