Ethyl isonicotinate ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Picolinic acid ethyl ester, 4-Pyridinecarboxylic acid ethyl ester, Pyridine-4-carboxylic acid ethyl ester, Ethyl 4-pyridinecarboxylate, Isonicotinic acid ethyl ester
136805-100G
136805-500G
BT136805-100GEA
57
USD
BT136805-100G
BT136805-500G
Ethyl isonicotinate ≥98%
Ethyl isonicotinate
CAS: 1570-45-2; EC No: 216-379-2; MDL No: MFCD00006428; RTECS: NS1450000
Liquid; Molecular Formula: C8H9NO2; MW: 151.17
Boiling Point: 92°/8 mmHg; Flash point: 88°C (190°F)
Density (g/mL): 1.009; Refractive Index: 1.501
- RTECS: NS1450000
- Refractive index: 1.501
- Liquid
Formula:
C₈H₉NO₂ Boiling Pt: 92 °C/8 mmHg Density: 1.009 g/mL Flash Pt: 88 °C (190 °F) |
MDL Number:
MFCD00006428 CAS Number: 1570-45-2 EINECS: 216-379-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...