Coenzyme Q10 ≥98%
Supplier: BeanTown Chemical
Synonyms:
(all-E)-2-(3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, Ubiquinone 10, Ubidecarenone
136760-1G
136760-25G
136760-5G
BT136760-1GEA
72.43
USD
BT136760-1G
BT136760-25G
BT136760-5G
Coenzyme Q10 ≥98%
Coenzyme Q10
CAS: 303-98-0; EC No: 206-147-9; MDL No: MFCD00042919; RTECS: DK3900000
Powder; Molecular Formula: C59H90O4; MW: 863.34
Light Sensitive
- RTECS: DK3900000
- Light Sensitive
- Powder
Formula:
C₅₉H₉₀O₄ MW: 863.37 g/mol |
MDL Number:
MFCD00042919 CAS Number: 303-98-0 EINECS: 206-147-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...