Zinc(II) acetylacetonate hydrate
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
Bis(2,4-pentanedionato-O,O')zinc (II) hydrate, Zinc acetylacetonate xhydrate, Zinc (II) 2,4-pentanedionate hydrate, Zinc (II) bis(2,4-pentanedionato-O,O') hydrate, Zinc acetylacetonate hydrate
136745-100G
136745-500G
BT136745-100GEA
74
USD
BT136745-100G
BT136745-500G
Zinc(II) acetylacetonate hydrate
Zinc(II) acetylacetonate hydrate
CAS: 14024-63-6; EC No: 237-860-3; MDL No: MFCD00149060; RTECS: ZH0917500
Solid; Linear Formula: Zn(C5H7O2)2·xH2O; MW: 263.61 (anhydrous)
Melting Point: 135-138°; Boiling Point: 129-131°/10 mmHg
- RTECS: ZH0917500
- Solid
Formula:
C₁₀H₁₄O₄Zn·H₂O MW: 281.62 Boiling Pt: 129…131 °C (10 mmHg) Melting Pt: 135…138 °C |
MDL Number:
MFCD00149060 CAS Number: 108503-47-5 EINECS: 237-860-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...