Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0)
Supplier: BeanTown Chemical
136720-1G
136720-5G
BT136720-1GEA
73
USD
BT136720-1G
BT136720-5G
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0)
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0)
CAS: 811862-77-8; MDL No: MFCD07369799
Solid; Molecular Formula: C42H44O10Pd; MW: 815.21
Melting Point: 170-178°
- Solid
Formula:
C₄₂H₄₄O₁₀Pd Melting Pt: 170…178 °C |
MDL Number:
MFCD07369799 CAS Number: 811862-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...