L-Valinamide hydrochloride ≥97%

Supplier: BeanTown Chemical

Synonyms: H-Val-NH2.HCl, H-Val-NH₂·HCl

136665-100G 136665-25G 136665-5G
BT136665-100GEA 519 USD
BT136665-100G BT136665-25G BT136665-5G
L-Valinamide hydrochloride ≥97%
L-Valinamide hydrochloride

CAS: 3014-80-0; MDL No: MFCD00039085 Solid; Molecular Formula: C5H12N2O·HCl; MW: 152.62 Melting Point: 266-270° Optical Rotation: [α]25/D +25.8°, c = 1 in water


  • Optical rotation: [α]25/D +25.8°, c = 1 in water
  • Solid

Formula: C₅H₁₂N₂O
MW: 152.62 g/mol
Melting Pt: 266…270 °C
MDL Number: MFCD00039085
CAS Number: 3014-80-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR