Tetradecylphosphonic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Tetradecylphosphonic acid, n-Tetradecylphosphonic acid, TDPA, N-tetradecylphosphonicacid

136650-1G 136650-25G 136650-5G
BT136650-1GEA 97 USD
BT136650-1G BT136650-25G BT136650-5G
Tetradecylphosphonic acid ≥98%
Tetradecylphosphonic acid

CAS: 4671-75-4; EC No: 225-118-1; MDL No: MFCD00015834 Solid; Linear Formula: CH3(CH2)13P(O)(OH)2; Molecular Formula: C14H31O3P; MW: 278.37 Melting Point: 80-85°


  • Solid

Formula: C₁₄H₃₁O₃P
MW: 278.37 g/mol
Melting Pt: 80…85 °C
MDL Number: MFCD00015834
CAS Number: 4671-75-4
EINECS: 225-118-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR