5-Hydroxy-2-adamantanone ≥98%
Supplier: BeanTown Chemical
136645-1G
136645-250MG
136645-5G
BT136645-1GEA
92
USD
BT136645-1G
BT136645-250MG
BT136645-5G
5-Hydroxy-2-adamantanone ≥98%
5-Hydroxy-2-adamantanone
CAS: 20098-14-0; MDL No: MFCD00192211
Solid; Molecular Formula: C10H14O2; MW: 166.22
Melting Point:
- Solid
Formula:
C₁₀H₁₄O₂ Melting Pt: >300 °C |
MDL Number:
MFCD00192211 CAS Number: 20098-14-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...