(3-Mercaptopropyl)triethoxysilane ≥94%
Supplier: BeanTown Chemical
Synonyms:
3-(Triethoxysilyl)propanethiol, 3-Mercaptopropyltriethoxysilane
136630-100G
136630-25G
136630-500G
BT136630-100GEA
120.04
USD
BT136630-100G
BT136630-25G
BT136630-500G
(3-Mercaptopropyl)triethoxysilane ≥94%
(3-Mercaptopropyl)triethoxysilane
CAS: 14814-09-6; EC No: 238-883-1; MDL No: MFCD00053924; RTECS: TZ7760000
Liquid; Molecular Formula: C9H22O3SSi; MW: 238.42
Flash point: 88°C (190°F)
Density (g/mL): 0.987; Refractive Index: 1.437
Air Sensitive, Moisture Sensitive
- RTECS: TZ7760000
- Refractive index: 1.437
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
HSH₂CCH₂CH₂Si(OCH₂CH₃)₃ MW: 238.42 g/mol Density: 0.987 g/mL Flash Pt: 88 °C (190 °F) |
MDL Number:
MFCD00053924 CAS Number: 14814-09-6 EINECS: 238-883-1 |
Specification Test Results
Assay | 94% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...