3-Nitro-p-toluic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Methyl-3-nitrobenzoic acid

136590-100G 136590-500G
BT136590-100GEA 46 USD
BT136590-100G BT136590-500G
3-Nitro-p-toluic acid ≥99%
3-Nitro-p-toluic acid

CAS: 96-98-0; EC No: 202-549-3; MDL No: MFCD00007174 Crystalline; Molecular Formula: C8H7NO4; MW: 181.15 Melting Point: 187-190°


  • Crystalline

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 187…190 °C
MDL Number: MFCD00007174
CAS Number: 96-98-0
EINECS: 202-549-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR