trans-4-Hydroxy-L(-)-proline ≥99%
Supplier: BeanTown Chemical
Synonyms:
L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
136565-100G
136565-25G
BT136565-100GEA
226.59
USD
BT136565-100G
BT136565-25G
trans-4-Hydroxy-L(-)-proline ≥99%
trans-4-Hydroxy-L(-)-proline
CAS: 51-35-4; EC No: 200-091-9; MDL No: MFCD00064320; RTECS: TW3586500
Crystalline; Molecular Formula: C5H9NO3; MW: 131.13
Melting Point: 273° (decomposes)
Optical Rotation: [α]25/D +75.6°, c = 1 in water
- RTECS: TW3586500
- Optical rotation: [α]25/D +75.6°, c = 1 in water
- Crystalline
Formula:
C₅H₉NO₃ Melting Pt: 273 °C (dec.) |
MDL Number:
MFCD00064320 CAS Number: 51-35-4 EINECS: 200-091-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...