Methyl-4-formylbenzoate ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl terephthalaldehydate, Methyl 4-formylbenzoate

136560-100G 136560-25G 136560-5G
BT136560-100GEA 752 USD
BT136560-100G BT136560-25G BT136560-5G
Methyl-4-formylbenzoate ≥98%
Methyl-4-formylbenzoate

CAS: 1571-08-0; EC No: 216-385-5; MDL No: MFCD00006950 Powder; Molecular Formula: C9H8O3; MW: 164.16 Melting Point: 59-63°; Boiling Point: 264-266°; Flash point: 128°C (262°F) Density (g/mL): 1.2


  • Powder

Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 264…266 °C
Melting Pt: 59…63 °C
Density: 1.2 g/mL
Flash Pt: 128 °C (262 °F)
MDL Number: MFCD00006950
CAS Number: 1571-08-0
EINECS: 216-385-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR