(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone ≥98%
Supplier: BeanTown Chemical
136470-1G
136470-5G
BT136470-1GEA
61
USD
BT136470-1G
BT136470-5G
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone ≥98%
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone
CAS: 17342-08-4; MDL No: MFCD00077792
Crystalline; Molecular Formula: C5H9NO2; MW: 115.13
Melting Point: 79-80°
Optical Rotation: [α]20/D +30°, c = 2 in ethanol
- Optical rotation: [α]20/D +30°, c = 2 in ethanol
- Crystalline
Formula:
C₅H₉NO₂ Melting Pt: 79…80 °C |
MDL Number:
MFCD00077792 CAS Number: 17342-08-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...