2-Chloro-4-nitropyridine ≥98%

Supplier: BeanTown Chemical

136465-1G 136465-5G
BT136465-1GEA 75 USD
BT136465-1G BT136465-5G
2-Chloro-4-nitropyridine ≥98%
2-Chloro-4-nitropyridine

CAS: 23056-36-2; MDL No: MFCD00661454 Solid; Molecular Formula: C5H3ClN2O2 ; MW: 158.54 Melting Point: 52-56°


  • Solid

Formula: C₅H₃ClN₂O₂
MW: 158.54
Melting Pt: 52…56 °C
MDL Number: MFCD00661454
CAS Number: 23056-36-2
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR