Thulium(III) oxide ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

136450-1G 136450-5G
BT136450-1GEA 111 USD
BT136450-1G BT136450-5G
Thulium(III) oxide ≥99.995% (trace metals basis)
Thulium(III) oxide

CAS: 12036-44-1; EC No: 234-851-6; MDL No: MFCD00011285 Powder; Linear Formula: Tm2O3; MW: 385.87 Melting Point: 2425° Hygroscopic


  • Hygroscopic
  • Powder

Formula: O₃Tm₂
MW: 385.87 g/mol
Melting Pt: 2425 °C
MDL Number: MFCD00011285
CAS Number: 12036-44-1
EINECS: 234-851-6

Order Now

Specification Test Results

Assay 99.995%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR