Iron ≥99.98% (trace metals basis), granules 1-2 mm
Supplier: BeanTown Chemical
136395-1KG
136395-250G
BT136395-1KGEA
100.03
USD
BT136395-1KG
BT136395-250G
Iron ≥99.98% (trace metals basis), granules 1-2 mm
Iron
CAS: 7439-89-6; EC No: 231-096-4; MDL No: MFCD00010999; RTECS: NO4565500
Solid; Molecular Formula: Fe; MW: 55.85
Melting Point: 1535°; Boiling Point: 2750°
Density (g/mL): 7.86
- RTECS: NO4565500
- Solid
Formula:
Fe Boiling Pt: 2750 °C Melting Pt: 1535 °C Density: 7.86 g/mL |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 |
Specification Test Results
Assay | 99.98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...