3,5-Dibromotoluene ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,3-Dibromo-5-methylbenzene

136350-100G 136350-25G 136350-5G
BT136350-100GEA 674 USD
BT136350-100G BT136350-25G BT136350-5G
3,5-Dibromotoluene ≥98%
3,5-Dibromotoluene

CAS: 1611-92-3; MDL No: MFCD00013528 Crystalline; Molecular Formula: C7H6Br2; MW: 249.93 Melting Point: 34-38°; Boiling Point: 246°; Flash point: 110°C (230°F)


  • Crystalline

Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 246 °C
Melting Pt: 34…38 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00013528
CAS Number: 1611-92-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR