(1R,3S)-(+)-Camphoric acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
D(+)-Camphoric acid, (1R,3S)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid, Levocarnitine Impurity B
136320-100G
136320-25G
136320-500G
BT136320-100GEA
80
USD
BT136320-100G
BT136320-25G
BT136320-500G
(1R,3S)-(+)-Camphoric acid ≥98%
(1R,3S)-(+)-Camphoric acid
CAS: 124-83-4; EC No: 204-715-0; MDL No: MFCD00001375
Crystalline; Molecular Formula: C10H16O4 ; MW: 200.23
Melting Point: 183-186°
Optical Rotation: [α]20/D +46°, c = 1 in ethanol
- Optical rotation: [α]20/D +46°, c = 1 in ethanol
- Crystalline
Formula:
C₁₀H₁₆O₄ MW: 200.23 g/mol Melting Pt: 183…186 °C |
MDL Number:
MFCD00001375 CAS Number: 124-83-4 EINECS: 204-715-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...