(4-Chlorophenoxy)acetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-CPA

136295-100G 136295-2.5KG 136295-500G
BT136295-100GEA 26.61 USD
BT136295-100G BT136295-2.5KG BT136295-500G
(4-Chlorophenoxy)acetic acid ≥98%
(4-Chlorophenoxy)acetic acid

CAS: 122-88-3; EC No: 204-581-3; MDL No: MFCD00004305; RTECS: AG0175000 Powder; Molecular Formula: C8H7ClO3; MW: 186.59 Melting Point: 157-161°


  • RTECS: AG0175000
  • Powder

Formula: (C₆H₄Cl)OCH₂COOH
MW: 186.59 g/mol
Melting Pt: 157…161 °C
MDL Number: MFCD00004305
CAS Number: 122-88-3
EINECS: 204-581-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR