Ethyl glycolate ≥95%
Supplier: BeanTown Chemical
Synonyms:
Ethyl Hydroxyacetate, Hydroxyacetic Acid Ethyl Ester, Glycolic Acid Ethyl Ester, Ethyl 2-hydroxyacetate
136270-10G
136270-50G
BT136270-10GEA
50
USD
BT136270-10G
BT136270-50G
Ethyl glycolate ≥95%
Ethyl glycolate
CAS: 623-50-7; EC No: 210-798-4; MDL No: MFCD00021970; RTECS: MC5410000
Liquid; Linear Formula: HOCH3CO2CH2CH3; Molecular Formula: C4H8O3; MW: 104.11
Boiling Point: 158-159°; Flash point: 62°C (144°F)
Density (g/mL): 1.1; Refractive Index: 1.419
- RTECS: MC5410000
- Refractive index: 1.419
- Liquid
Formula:
C₄H₈O₃ Boiling Pt: 158…159 °C Density: 1.1 g/mL Flash Pt: 62 °C (144 °F) |
MDL Number:
MFCD00021970 CAS Number: 623-50-7 EINECS: 210-798-4 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...