3-Amino-4-formylpyridine ≥95%

Supplier: BeanTown Chemical

Synonyms: 3-Amino-pyridine-4-carbaldehyde, 3-Aminoisonicotinaldehyde, 3-Aminopyridine-4-carboxaldehyde

136250-1G 136250-250MG
BT136250-1GEA 266.12 USD
BT136250-1G BT136250-250MG
3-Amino-4-formylpyridine ≥95%
3-Amino-4-formylpyridine

CAS: 55279-29-3; MDL No: MFCD06410684 Solid; Molecular Formula: C6H6N2O; MW: 122.12 Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₆H₆N₂O
MW: 122.13 g/mol
MDL Number: MFCD06410684
CAS Number: 55279-29-3

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR