3-Nitrophthalic anhydride ≥97%
Supplier: BeanTown Chemical
136240-25G
136240-500G
BT136240-100GEA
533.61
USD
BT136240-100G
BT136240-25G
BT136240-500G
3-Nitrophthalic anhydride ≥97%
3-Nitrophthalic anhydride
CAS: 641-70-3; EC No: 211-373-6; MDL No: MFCD00005921
Crystalline/Powder; Molecular Formula: C8H3NO5 ; MW: 193.11
Melting Point: 163-165°
Moisture Sensitive
- Moisture Sensitive
- Crystalline/Powder
Formula:
C₈H₃NO₅ Melting Pt: 163…165 °C |
MDL Number:
MFCD00005921 CAS Number: 641-70-3 EINECS: 211-373-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...