6-Nitro-4-quinazolone ≥96%

Supplier: BeanTown Chemical

Synonyms: 6-Nitroquinazolin-4(3H)-one, 6-Nitro-4(3H)-quinazolinone

136235-1G 136235-5G
BT136235-1GEA 171 USD
BT136235-1G BT136235-5G
6-Nitro-4-quinazolone ≥96%
6-Nitro-4-quinazolone

CAS: 6943-17-5 Solid; Molecular Formula: C8H5N3O3; MW: 191.14


  • Solid

Formula: C₈H₅N₃O₃
MW: 191.1436 g/mol
MDL Number: MFCD00137845
CAS Number: 6943-17-5

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR