1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS ≥95%
Supplier: BeanTown Chemical
Synonyms:
CTC-Q-PHOS, 1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene, QPhos, 1,2,3,4,5-Pentaphenyl-1'-(di-t-butylphosphino)ferrocene
136185-250MG
136185-500MG
BT136185-250MGEA
200
USD
BT136185-250MG
BT136185-500MG
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS ≥95%
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS
CAS: 312959-24-3; MDL No: MFCD03788940
Powder; Molecular Formula: C48H47FeP; MW: 710.71
Melting Point: 211-219°
- Powder
Formula:
C₄₈H₄₇FeP MW: 710.706 g/mol Melting Pt: 211…219 °C |
MDL Number:
MFCD03788940 CAS Number: 312959-24-3 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...