4-Iodo-o-xylene ≥98%

Supplier: BeanTown Chemical

Synonyms: 3,4-Dimethyliodobenzene, 1,2-Dimethyl-4-iodobenzene, 3,4-Dimethyl-1-iodobenzene, 4-Iodo-1,2-dimethylbenzene, 1-Iodo-3,4-dimethylbenzene

136160-10G 136160-250G 136160-50G
BT136160-10GEA 49 USD
BT136160-10G BT136160-250G BT136160-50G
4-Iodo-o-xylene ≥98%
4-Iodo-o-xylene

CAS: 31599-61-8; MDL No: MFCD00040989 Liquid; Molecular Formula: C8H9I; MW: 232.07 Boiling Point: 106-108°/13 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.633; Refractive Index: 1.603 Light Sensitive


  • Refractive index: 1.603
  • Light Sensitive
  • Liquid

Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 106…108 °C/13 mmHg
Density: 1.633 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00040989
CAS Number: 31599-61-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR