3-Aminoquinoline ≥98%

Supplier: BeanTown Chemical

Synonyms: Quinolin-3-amine, Quinolin-3-ylamine

136150-1G 136150-25G 136150-5G
BT136150-1GEA 21.6 USD
BT136150-1G BT136150-25G BT136150-5G
3-Aminoquinoline ≥98%
3-Aminoquinoline

CAS: 580-17-6; EC No: 209-455-1; MDL No: MFCD00006772; RTECS: VA9622000 Crystalline; Molecular Formula: C9H8N2 ; MW: 144.17 Melting Point: 91-92° Air Sensitive


  • RTECS: VA9622000
  • Air Sensitive
  • Crystalline

Formula: C₉H₈N₂
MW: 144.18 g/mol
Melting Pt: 91…92 °C
MDL Number: MFCD00006772
CAS Number: 580-17-6
EINECS: 209-455-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR