4-tert-Butylcyclohexanone ≥99%
Supplier: BeanTown Chemical
136140-100G
136140-500G
BT136140-100GEA
57
USD
BT136140-100G
BT136140-500G
4-tert-Butylcyclohexanone ≥99%
4-tert-Butylcyclohexanone
CAS: 98-53-3; EC No: 202-678-5; MDL No: MFCD00001642; RTECS: GW1140000
Crystalline; Molecular Formula: C10H18O; MW: 154.25
Melting Point: 47-50°; Boiling Point: 113-116°/20 mmHg; Flash point: 96°C (205°F)
- RTECS: GW1140000
- Crystalline
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 113…116 °C/20 mmHg Melting Pt: 47…50 °C Flash Pt: 96 °C (205 °F) |
MDL Number:
MFCD00001642 CAS Number: 98-53-3 EINECS: 202-678-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...