Iron(II) oxalate dihydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Iron (II) ethanedioate dihydrate, Oxalic acid iron (II) salt dihydrate, Ferrous oxalate dihydrate, Iron(II) oxalate hydrate
136120-1KG
136120-250G
BT136120-1KGEA
153
USD
BT136120-1KG
BT136120-250G
Iron(II) oxalate dihydrate ≥99%
Iron(II) oxalate dihydrate
CAS: 6047-25-2; EC No: 208-217-4; MDL No: MFCD00150040
Powder; Linear Formula: FeC2O4·2H2O; MW: 179.90
Density (g/mL): 2.28
Hygroscopic
- Hygroscopic
- Powder
Formula:
Fe(C₂O₄)·2H₂O MW: 179.9 g/mol Density: 2.28 g/mL |
MDL Number:
MFCD00150040 CAS Number: 6047-25-2 EINECS: 208-217-4 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...