4,5,6-Triaminopyrimidine sulfate hydrate, (max. 8% H₂O) ≥98%

Supplier: BeanTown Chemical

136110-1G 136110-5G
BT136110-1GEA 38 USD
BT136110-1G BT136110-5G
4,5,6-Triaminopyrimidine sulfate hydrate, (max. 8% H₂O) ≥98%
4,5,6-Triaminopyrimidine sulfate hydrate

CAS: 207742-76-5; EC No: 256-446-3; MDL No: MFCD00012789 Powder; Molecular Formula: C4H7N5·H2SO4·xH2O; MW: 223.22 (anhydrous) Melting Point: 300°


  • Powder

Formula: C₄H₇N₅·H₂SO₄·xH₂O
MW: 223.22 g/mol (anhy)
Melting Pt: >300 °C
MDL Number: MFCD00012789
CAS Number: 207742-76-5
EINECS: 256-446-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR