Tris(perfluorobutyl)amine ≥90%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

136085-25G 136085-5G
BT136085-25GEA 107 USD
BT136085-25G BT136085-5G
Tris(perfluorobutyl)amine ≥90%, Technical Grade
Tris(perfluorobutyl)amine

CAS: 311-89-7; EC No: 206-223-1; MDL No: MFCD00000436; RTECS: YA1000000 Liquid; Linear Formula: [CF3(CF2)3]3N; Molecular Formula: C12F27N; MW: 671.09 Melting Point: -52°; Boiling Point: 177-180° Density (g/mL): 1.883; Refractive Index: 1.289


  • RTECS: YA1000000
  • Refractive index: 1.289
  • Liquid

Formula: C₁₂F₂₇N
MW: 671.1 g/mol
Boiling Pt: 177…180 °C
Melting Pt: –52 °C
Density: 1.883 g/mL
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR