Diyttrium trioxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Yttrium oxide, Yttrium (III) oxide

136080-1KG 136080-250G 136080-50G
BT136080-1KGEA 736 USD
BT136080-1KG BT136080-250G BT136080-50G
Diyttrium trioxide ≥99.9%
Diyttrium trioxide

CAS: 1314-36-9; EC No: 215-233-5; MDL No: MFCD00011473 10 Micron Powder; Linear Formula: Y2O3; MW: 225.81 Melting Point: 2410°; Boiling Point: 4300° Density (g/mL): 5.01 Air Sensitive


  • Air Sensitive
  • <10 Micron Powder

Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300
Melting Pt: 2410
Density: 5.01 g/mL
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR