PTFE

Supplier: BeanTown Chemical

136055-1KG 136055-250G 136055-50G
BT136055-1KGEA 564 USD
BT136055-1KG BT136055-250G BT136055-50G
PTFE
PTFE

CAS: 9002-84-0; MDL No: MFCD00084455; RTECS: KX4025000 Solid; Linear Formula: (-CF2CF2-)n Melting Point: 321° Density (g/mL): 2


  • RTECS: KX4025000
  • Solid

Formula: (CF₂CF₂)n
Melting Pt: 321 °C
Density: 2 g/mL
MDL Number: MFCD00084455
CAS Number: 9002-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR