Hexadecan-16-olide ≥97%
Supplier: BeanTown Chemical
136035-10G
136035-50G
BT136035-10GEA
76
USD
BT136035-10G
BT136035-50G
Hexadecan-16-olide ≥97%
Hexadecan-16-olide
CAS: 109-29-5; EC No: 203-662-0; MDL No: MFCD00039668; RTECS: RN9730000
Solid; Molecular Formula: C16H30O2; MW: 254.41
Melting Point: 34-38°
- RTECS: RN9730000
- Solid
Formula:
C₁₆H₃₀O₂ Melting Pt: 34…38 °C |
MDL Number:
MFCD00039668 CAS Number: 109-29-5 EINECS: 203-662-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...