1,3-Diphenylacetone ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone
135995-100G
135995-25G
BT135995-100GEA
167.5
USD
BT135995-100G
BT135995-25G
1,3-Diphenylacetone ≥98%
1,3-Diphenylacetone
CAS: 102-04-5; EC No: 203-000-0; MDL No: MFCD00004795; RTECS: UC2010030
Solid; Molecular Formula: C15H14O; MW: 210.27
Melting Point: 32-34°; Boiling Point: 330°; Flash point:
- RTECS: UC2010030
- Solid
Formula:
(C₆H₅)CH₂C(O)CH₂(C₆H₅) MW: 210.28 g/mol Boiling Pt: 330 °C Melting Pt: 32…34 °C Density: 1.04 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00004795 CAS Number: 102-04-5 EINECS: 203-000-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...