4,6-Dichloro-5-pyrimidinecarboxylic acid ≥95%

Supplier: BeanTown Chemical

135985-1G 135985-250MG 135985-5G
BT135985-1GEA 70 USD
BT135985-1G BT135985-250MG BT135985-5G
4,6-Dichloro-5-pyrimidinecarboxylic acid ≥95%
4,6-Dichloro-5-pyrimidinecarboxylic acid

CAS: 87600-98-4 Solid; Molecular Formula: C5H2Cl2N2O2; MW: 192.99


  • Solid

Formula: C₅H₂Cl₂N₂O₂
MDL Number: MFCD09909611
CAS Number: 87600-98-4

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR