7-Bromoquinazolin-4(3H)-one ≥97%

Supplier: BeanTown Chemical

Synonyms: 7-Bromo-3,4-dihydroquinazolin-4-one

135930-1G 135930-5G
BT135930-1GEA 117 USD
BT135930-1G BT135930-5G
7-Bromoquinazolin-4(3H)-one ≥97%
7-Bromoquinazolin-4(3H)-one

CAS: 194851-16-6 Solid; Molecular Formula: C8H5BrN2O; MW: 225.04 Melting Point: 262-267°


  • Solid

Formula: C₈H₅BrN₂O
MW: 225.0400 g/mol
Melting Pt: 262…267 °C
MDL Number: MFCD07838323
CAS Number: 194851-16-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR