5-Hydroxy-DL-tryptophan ≥99%
Supplier: BeanTown Chemical
Synonyms:
DL-5-Hydroxytryptophan, 2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
135925-1G
135925-5G
BT135925-1GEA
59.81
USD
BT135925-1G
BT135925-5G
5-Hydroxy-DL-tryptophan ≥99%
5-Hydroxy-DL-tryptophan
CAS: 56-69-9; EC No: 200-284-8; MDL No: MFCD00005651; RTECS: YN7100000
Solid; Molecular Formula: C11H12N2O3; MW: 220.22
Melting Point: 298-300°
Density (g/mL): 1.484
- RTECS: YN7100000
- Solid
Formula:
C₁₁H₁₂N₂O₃ MW: 220.23 g/mol Melting Pt: 298…300 °C Density: 1.484 g/mL |
MDL Number:
MFCD00005651 CAS Number: 56-69-9 EINECS: 200-284-8 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...