4-Bromo-2,6-difluorophenol ≥97%
Supplier: BeanTown Chemical
135905-1G
135905-5G
BT135905-1GEA
55
USD
BT135905-1G
BT135905-5G
4-Bromo-2,6-difluorophenol ≥97%
4-Bromo-2,6-difluorophenol
CAS: 104197-13-9; MDL No: MFCD00236213
Solid; Molecular Formula: C6H3BrF2O; MW: 208.99
Melting Point: 50°
- Solid
Formula:
C₆H₃BrF₂O Melting Pt: 50 °C |
MDL Number:
MFCD00236213 CAS Number: 104197-13-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...