Calcium aluminate ≥99%

Supplier: BeanTown Chemical

135885-100G 135885-25G 135885-5G
BT135885-100GEA 426 USD
BT135885-100G BT135885-25G BT135885-5G
Calcium aluminate ≥99%
Calcium aluminate

CAS: 12042-68-1; EC No: 234-931-0; MDL No: MFCD00049722 -200 Mesh Powder; Linear Formula: CaAl2O4; MW: 158.02 Hygroscopic


  • Hygroscopic
  • -200 Mesh Powder

Formula: Al₂CaO₄
MW: 158.04 g/mol
MDL Number: MFCD00049722
CAS Number: 12042-68-1
EINECS: 234-931-0

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR