Palladium ≥99.9% (trace metals basis), foil, Thickness 0.1 mm

Supplier: BeanTown Chemical

135850-100X100 135850-25X25MM
BT135850-100X1EA 2155 USD
BT135850-100X1 BT135850-25X25
Palladium ≥99.9% (trace metals basis), foil, Thickness 0.1 mm
Palladium

CAS: 7440-05-3; EC No: 231-115-6; MDL No: MFCD00011167; RTECS: RT3480500 Solid; Molecular Formula: Pd; MW: 106.42 Melting Point: 1554°; Boiling Point: 2970° Density (g/mL): 12.02


  • RTECS: RT3480500
  • Solid

Formula: Pd
MW: 106.420 g/mol
Boiling Pt: 2970 °C
Melting Pt: 1554 °C
Density: 12.02 g/mL
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR