(R)-(+)-4-Benzyl-2-oxazolidinone ≥99%

Supplier: BeanTown Chemical

Synonyms: (R)-4-Benzyl-2-oxazolidinone

135840-1G 135840-25G 135840-5G
BT135840-1GEA 44 USD
BT135840-1G BT135840-25G BT135840-5G
(R)-(+)-4-Benzyl-2-oxazolidinone ≥99%
(R)-(+)-4-Benzyl-2-oxazolidinone

CAS: 102029-44-7; MDL No: MFCD00010846 Crystalline/Powder; Molecular Formula: C10H11NO2 ; MW: 177.20 Melting Point: 88-90° Optical Rotation: [α]18/D +64°, c = 1 in chloroform Hygroscopic


  • Optical rotation: [α]18/D +64°, c = 1 in chloroform
  • Hygroscopic
  • Crystalline/Powder

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 88…90 °C
MDL Number: MFCD00010846
CAS Number: 102029-44-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR