Cholic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxycholan-24-oic acid, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,7α,12α-Trihydroxycholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid
135810-100G
135810-25G
135810-500G
BT135810-100GEA
121.73
USD
BT135810-100G
BT135810-25G
BT135810-500G
Cholic acid ≥98%
Cholic acid
CAS: 81-25-4; EC No: 201-337-8; MDL No: MFCD00003672; RTECS: FZ9350000
Powder; Molecular Formula: C24H40O5; MW: 408.57
Melting Point: 200-201°
- RTECS: FZ9350000
- Powder
Formula:
C₂₄H₄₀O₅ MW: 408.58 g/mol Melting Pt: 200…201 °C |
MDL Number:
MFCD00003672 CAS Number: 81-25-4 EINECS: 201-337-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...